missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic acid 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | L-Ascorbic Acid |
| CAS | 50-81-7 |
| Analyte or Component Names | L-Ascorbic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H8 O6 |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | L-Ascorbic Acid, (+)-Ascorbic acid, 100M, 3-Oxo-L-gulofuranolactone, 3-keto-L-Gulofuranolactone, Adenex, Allercorb, Antiscorbic vitamin, Antiscorbutic vitamin, Ascoltin, Ascorbajen, Ascorbic acid, Ascorbicap, Ascorbutina, Ascorell, Ascorin, Ascorteal, Ascorvit, C-L 6/PW, C-Quin, C-Vimin, Cantan, Cantaxin, Catavin C, Ce-Mi-Lin, Ce-Vi-Sol, Cebicure, Cebion, Cebione, Cecon, Cegiolan, Ceglion, Ceklin, Celaskon, Celin, Cell C, Cemagyl, Cenetone, Cereon, Cergona, Cescorbat, Cetamid, Cetane, Cetane-Caps TC, Cetebe, Cetemican, Cevalin, Cevarol, Cevatine, Cevex, Cevimin, Cevital, Cevitamic acid, Cevitamin, Cevitan, Cevitex, Cewin, Chewcee, Ciamin, Cinal, Cipca, Citrovit, Colascor, Concemin, Davitamon C, E 300, Hicee, Hybrin, IDO-C, Juvamine, Kangbingfeng, L-(+)-Ascorbic acid, L-3-Keto-threo-hexuronic acid lactone, L-Lyxoascorbic acid, L-Xyloascorbic acid, L-threo-Ascorbic acid, Laroscorbine, Lemascorb, Liqui-Cee, NSC 218455, NSC 33832, Neo-Valdrin, P 1110, Planavit C, Proscorbin, Redoxon, Ribena, Ronotec 100, Rontex 100, Rovimix C, Rovimix C-EC, Scorbu C, Secorbate, Suncoat VC 40, Testascorbic, VC 40, VC 97, Vasc, Vicelat, Vicin, Viforcit, Viscorin, Viscorin 100M, Vitace, Vitacee, Vitacimin, Vitacin, Vitamin C, Vitamisin, Vitascorbol, Vitashure 160, Xitix, L-threo-Hex-2-enonic acid, gamma-lactone |
| InChI Formula | InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1 |
| SMILES | OC[C@H](O)[C@H]1OC(=O)C(=C1O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?