missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, TRC

Product Code. 17978864
Change view
Click to view available options
Quantity:
100 mg
25 g
Unit Size:
100mg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17978864 25 g 25g
17928564 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17978864 Supplier Toronto Research Chemicals Supplier No. TRCA76950525G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6 H14 N4 O2
Molecular Weight (g/mol) 174.2
Synonym Arginine, L- (8CI), (S)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid, A 0526, Arginine, L-(+)-Arginine, L-Arg, L-Norvaline, 5-[(aminoiminomethyl)amino]-, L-Ornithine, N5-(aminoiminomethyl)-, L-arginine C Grade, L-α-Amino-δ-guanidinovaleric acid, NSC 206269, Pentanoic acid, 2-amino-5-[(aminoiminomethyl)amino]-, (S)-, L-Arginine
IUPAC Name (2S)-2-amino-5-carbamimidamidopentanoic acid
SMILES N[C@@H](CCCNC(=N)N)C(=O)O

Specifications

Analyte or Component Names L-Arginine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 25 g
Formula Weight 174.1117
Percent Purity >95%
Chemical Name or Material L-Arginine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.