missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine methyl ester dihydrochloride, 98%

Product Code. 11362306
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362306 25 g 25g
11352306 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362306 Supplier Thermo Scientific Alfa Aesar Supplier No. L05959.14
  • ONLINE EXCLUSIVE PRICE
    £100.06
    £88.65 / 25g
    Save £11.41
    11% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26340-89-6 | C7H18Cl2N4O2 | 261.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26340-89-6
Molecular Formula C7H18Cl2N4O2
Molecular Weight (g/mol) 261.147
MDL Number MFCD00038948
InChI Key XQYZOBNLCUAXLF-XRIGFGBMSA-N
PubChem CID 117776
IUPAC Name methyl (2S)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
SMILES COC(=O)C(CCCN=C(N)N)N.Cl.Cl

Specifications

Melting Point ∽190°C (decomposition)
Beilstein 4159929
Synonym l-arginine methyl ester dihydrochloride, h-arg-ome.2hcl, methyl l-argininate dihydrochloride, h-arg-ome 2hcl, l-arginine, methyl ester, dihydrochloride, s-methyl 2-amino-5-guanidinopentanoate dihydrochloride, methyl 2s-2-amino-5-carbamimidamidopentanoate dihydrochloride, l-arginine, methyl ester, hydrochloride 1:2, h-arg-ome?hcl, h-arg-ome 2 hcl
Quantity 25 g
Formula Weight 261.15
Optical Rotation +21° (c=2.5 in Methanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Arginine methyl ester dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.