missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine Hydrochloride, TRC

Product Code. 30335938
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335938 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335938 Supplier Toronto Research Chemicals Supplier No. TRCA769500100G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arginine Hydrochloride
CAS 1119-34-2
Molecular Formula C6 H14 N4 O2 . Cl H
Quantity 100 g
Synonym Arginine Hydrochloride, L-Arginine Monohydrochloride, L-Arginine Hydrochloride, Argamine, Argivene, Detoxargin, L-(+)-Arginine Monohydrochloride, Levargin, Minophagen A, NSC 203450, NSC 7914, R-Gene
InChI Formula InChI=1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1
SMILES Cl.N[C@@H](CCCNC(=N)N)C(=O)O
IUPAC Name (2S)-2-amino-5-carbamimidamidopentanoic acid;hydrochloride
Molecular Weight (g/mol) 210.66
Formula Weight 210.0884
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.