missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Hydrochloride, MP Biomedicals™

Product Code. 11421605 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421605 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421605 Supplier MP Biomedicals Supplier No. 0210074396
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Necessary for the production of protein, helps rid the body of ammonia and stimulates insulin release. L-Arginine, Hydrochloride, MP Biomedicals is an amino acid and substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO

L-arginine is necessary for the production of protein. It also helps rid the body of ammonia (a waste product). It stimulates the release of insulin by a nitric oxide-dependent mechanism. In addition, L-arginine is used to make nitric oxide (a compound that relaxes the blood vessels).

Specifications

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
Solubility Information Soluble in water (50mg/mL); slightly soluble in hot alcohol; insoluble in ether.
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
Molecular Weight (g/mol) 210.662
Quantity 2.5 kg
PubChem CID 66250
Formula Weight 210.67
Percent Purity 99.6%
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine Monohydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.