missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine hydrochloride, MedChemExpress

Product Code. 16482529
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482529 1 g 1g
16431418 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482529 Supplier Medchem Express Supplier No. HYN0455A
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is the nitrogen donor for synthesis of nitric oxide, a potent vasodilator that is deficient during times of sickle cell crisis.

L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is the nitrogen donor for synthesis of nitric oxide, a potent vasodilator that is deficient during times of sickle cell crisis.

Specifications

Chemical Name or Material L-Arginine hydrochloride
CAS 1119-34-2
Color White
Purity Grade Notes Research
Molecular Formula C6H15ClN4O2
Synonym (S)-(+)-Arginine hydrochloride
Solubility Information H2O : 100 mg/mL (474.70 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (4.75 mM; ultrasonic and warming and heat to 80°C)
SMILES N[C@@H](CCCNC(N)=N)C(O)=O.[H]Cl
Molecular Weight (g/mol) 210.66
Quantity 1 g
Formula Weight 210.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.