missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Arginine HCl

Product Code. 30018045
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
Product Code. Quantity unitSize
30018045 100 g 100g
30018048 5 kg 5kg
30018046 1 kg 1kg
30018047 500 g 500g
4 options
This item is not returnable. View return policy

Product Code. 30018045

Brand: Melford A50400100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • CAS Number: 1119-34-2
  • Molecular Formula: C6H14N4O2 HCl
  • Molecular Weight: 210.7

Specifications

Chemical Name or Material L-Arginine HCl
CAS 1119-34-2
Molecular Formula C6H14N4O2-HCl
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.