missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Free Base, Fisher BioReagents

Product Code. 10421725
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
L-Arginine is an essential amino acid used in biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Melting Point 223°C
pH 11.4
Color White
Assay Percent Range ≥99 %
Identification Pass Test
Infrared Spectrum Conforms to reference
Loss on Drying 0.3% max.
Merck Index 15, 770
Packaging Poly Bottle
Specific Rotation +26.3 to +27.7° (+ or -)
Formula Weight 174.12
Percent Purity ≥99%
Quantity 500 g
Physical Form Solid
Chemical Name or Material L-Arginine, Free Base
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Arginine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'