missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Free Base, Fisher BioReagents

Product Code. 10368364
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10368364 1 kg Poly Bottle 1kg
10041693 100 g Poly Bottle 100g
10421725 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10368364 Supplier Fisher BioReagents Supplier No. BP25051
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
L-Arginine is an essential amino acid used in biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Melting Point 223°C
pH 11.4
Color White
Identification Pass Test
Infrared Spectrum Conforms to reference
Assay Percent Range ≥99 %
Loss on Drying 0.3% max.
Packaging Poly Bottle
Specific Rotation +26.3 to +27.7° (+ or -)
Merck Index 15, 770
Quantity 1 kg
Formula Weight 174.12
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material L-Arginine, Free Base
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Arginine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.