missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Extra Pure, SLR, Fisher Chemical

Product Code. 10122090 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10122090 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10122090 Supplier Fisher Chemical Supplier No. A/8300/46
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 223°C
CAS 74-79-3
pH 10.5 to 12
Color White
Molecular Formula C6H14N4O2
Packaging Plastic Powder Jar
MDL Number 2635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
SMILES NC(CCCN=C(N)N)C(O)=O
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol) 174.20
Quantity 25 g
Formula Weight 174.2g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Arginine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.