missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, 99%, [Free Base], MP Biomedicals

Product Code. 11445072 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445072 500 g 500g
11425072 25 g 25g
11435072 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445072 Supplier MP Biomedicals Supplier No. 0219462690
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74-79-3 | C6H14N4O2 | 174.20 g/mol

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Quantity 500 g
Formula Weight 174.2
Percent Purity 100%
Grade Cell Culture
Chemical Name or Material L-Arginine, 99%, Free Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.