missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Arabitol, 98%

Product Code. 11454444
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454444 100 g 100g
11434444 5 g 5g
11444444 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454444 Supplier Thermo Scientific Chemicals Supplier No. A13103.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7643-75-6 | C5H12O5 | 152.15 g/mol

L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

Solubility
Soluble in water (50 mg/ml).

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7643-75-6
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064290
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
PubChem CID 439255
ChEBI CHEBI:18403
IUPAC Name (2S,4S)-pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 98°C to 103°C
Quantity 100 g
Beilstein 1720521
Sensitivity Hygroscopic
Merck Index 14,762
Solubility Information Soluble in water (50mg/ml).
Optical Rotation −11° (c=5 in 8% Borax Solution)
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material L-(-)-Arabitol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.