missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Arabinose, MP Biomedicals™

Product Code. 11411605 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411605 250 g 250g
11401605 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411605 Supplier MP Biomedicals Supplier No. 0210070683
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a substrate to identify, differentiate and characterize pentose sugar isomerases. L-(+)-Arabinose, MP Biomedicals is the naturally occurring isomer and is a constituent of plant polysaccharides. It is used in the bioproduction of L-ribose

Most bacteria contain an inducible arabinose operon that codes for a series of enzymes and transporters that allows L-arabinose to be used as the sole carbon source in microbial culture.

Chemical Identifiers

CAS 5328-37-0
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key PYMYPHUHKUWMLA-VAYJURFESA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 152°C to 165°C
Density 1.585g/mL (20°C) (Lit.)
Quantity 250 g
Solubility Information Freely soluble in water; Partially soluble in ethanol 90%.
Optical Rotation (c=1, water) 100–110°
Formula Weight 150.1
Percent Purity ≥98%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.