missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Arabinose, MedChemExpress

Product Code. 16463449
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463449 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16463449 Supplier Medchem Express Supplier No. HYW015611
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-(+)-Arabinose selectively inhibits intestinal sucrase activity in a noncompetitive manner and suppresses the plasma glucose increase due to sucrose ingestion.

L-(+)-Arabinose selectively inhibits intestinal sucrase activity in a noncompetitive manner and suppresses the plasma glucose increase due to sucrose ingestion.

Specifications

Chemical Name or Material L-(+)-Arabinose
CAS 5328-37-0
Color White
Purity Grade Notes Research
Molecular Formula C5H10O5
Quantity 1 g
Solubility Information H2O : 500 mg/mL (3330.45 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (333.04 mM; Need ultrasonic)
SMILES O=C[C@@H]([C@H]([C@H](CO)O)O)O
Molecular Weight (g/mol) 150.13
Formula Weight 150.13
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.