missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Arabinose, 99%, for biochemistry

Product Code. 15209295 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15209295 500 g 500g
15289285 25 g 25g
15299285 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15209295 Supplier Thermo Scientific Chemicals Supplier No. 365185000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-72-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-VVZXFQNISA-N
Synonym beta-d-arabinopyranose, 2r,3s,4r,5r-oxane-2,3,4,5-tetrol, pentopyranose #, .beta.-d-arabinopyranose, b-arabinopyranose, beta-arabinopyranose 9ci, methyl, a-d-arabinopyranoside, beta-d-arabinopyranose 9ci, 2r,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol, 3s,2r,4r,5r-2h-3,4,5,6-tetrahydropyran-2,3,4,5-tetraol
PubChem CID 25245970
IUPAC Name (4S,5S)-oxane-2,3,4,5-tetrol
SMILES C1C(C(C(C(O1)O)O)O)O

Specifications

Melting Point 155.0°C to 160.0°C
Ignition Residue 0.3% max
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Quantity 500 g
Beilstein 31,32
Merck Index 10,783
Specific Rotation + 103.00
Solubility Information (10% in water) Clear colorless
Formula Weight 150.13
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material L(+)-Arabinose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.