missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Arabinose, ≥98%, MP Biomedicals™

Product Code. 11417800 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417800 100 g 100g
11427800 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417800 Supplier MP Biomedicals Supplier No. 0210070680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5328-37-0
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key PYMYPHUHKUWMLA-VAYJURFESA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Color White
Density 1.585g/mL at 20°C
Assay Percent Range ≥98%
Quantity 100 g
Formula Weight 150.1
Physical Form Powder
Chemical Name or Material L-(+)-Arabinose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.