missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-AP4

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
Selective group III metabotropic glutamate receptor agonist. Synaptic depressant. Agonist at the quisqualate-sensitized AP6 site in hippocampus.
Available as part of the Group III mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . DL Mixture and D-isomer also available.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 183.1 |
| Purity | >99% |
| Molecular Weight (g/mol) | 183.1 |
| Chemical Name or Material | L-AP4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction