missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Aminobutyric Acid, TRC
Specifications
Specifications
| CAS | 1492-24-6 |
| Analyte or Component Names | L-Aminobutyric Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H9 N O2 |
| Synonym | Butanoic acid, 2-amino-, (2S)-, Butanoic acid, 2-amino-, (S)-, Butyric acid, 2-amino-, L- (8CI), (2S)-2-Aminobutanoic acid, (+)-2-Aminobutanoic acid, (+)-2-Aminobutyric acid, (+)-α-Aminobutyric acid, (S)-(+)-α-Aminobutyric acid, (S)-2-Aminobutanoic acid, (S)-2-Aminobutyric acid, L-2-Amino-n-butyric acid, L-2-Aminobutanoic acid, L-2-Aminobutyric acid, L-Butyrine, L-Ethylglycine, L-Homoalanine, L-α-Amino-n-butyric acid, L-α-Aminobutyric acid, NSC 97060, S-Butyrine |
| InChI Formula | InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC[C@H](N)C(=O)O |
| IUPAC Name | (2S)-2-aminobutanoic acid |
| Industry Type | Forensics |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?