Learn More
Abcam L-Amino Acid Assay Kit
Product Code. 32173965 Shop All Abcam Products
Change view
Click to view available options
100 Tests
Product Code.
32173965 100 Tests
1 options
Product Code. 32173965 Supplier Abcam Supplier No. ab65347100Test
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Assay L-Amino Acid Assay in 30m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with L-Amino Acid Assay Kit AB65347. Sensitivity ≤ 40 μM. Cited in 26 publications. Use with Microplate reader.
Assay L-Amino Acid Assay in 30m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with L-Amino Acid Assay Kit AB65347. Sensitivity ≤ 40 μM. Cited in 26 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests