missing translation for 'onlineSavingsMsg'
Learn More

L-Alpha-Phosphatidyl-L-serine Sodium Salt (Technical Grade), TRC

Product Code. 30012616
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012616 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012616 Supplier Toronto Research Chemicals Supplier No. TRCP3537385MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 321595-13-5
Molecular Formula C42H81NNaO10P
Quantity 5 mg
Synonym (2R)-2,3-bis[(1-oxooctadecyl)oxy]propyl Hydrogen Phosphate (Ester) L-Serine Sodium Salt, (R)-2,3-Bis[(1-oxooctadecyl)oxy]propyl Hydrogen Phosphate (Ester) L-Serine Sodium Salt, 1,2-Distearoyl phosphatidyl Serine Sodium Salt, 1,2-Distearoyl-sn-3-glycerophosphoserine Sodium Salt, 1,2-Distearoyl-sn-glycero-3-phosphatidylserine Sodium Salt, DSPS Sodium Salt, Distearoylphosphatidylserine Sodium Salt
InChI Formula InChI=1S/C42H82NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(44)50-35-38(36-51-54(48,49)52-37-39(43)42(46)47)53-41(45)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h38-39H,3-37,43H2,1-2H3,(H,46,47)(H,48,49);/q;+1/p-1/t38-,39+;/m1./s1
SMILES [Na+].CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OC[C@H](N)C(=O)O)OC(=O)CCCCCCCCCCCCCCCCC
IUPAC Name sodium;[(2S)-2-amino-2-carboxyethyl] [(2R)-2,3-di(octadecanoyloxy)propyl] phosphate
Molecular Weight (g/mol) 814.07
Formula Weight 813.5496
Chemical Name or Material L-alpha-Phosphatidyl-L-serine Sodium Salt (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.