missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide, 98%, Thermo Scientific™

Product Code. 11456376
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456376 0.25 g 0.25g
11446376 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456376 Supplier Thermo Scientific Alfa Aesar Supplier No. B23148.01

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15295-77-9
Molecular Formula C4H8BrNO2
Molecular Weight (g/mol) 182.02
MDL Number MFCD00012723,MFCD00674493
InChI Key MKLNTBLOABOJFZ-UHFFFAOYNA-N
Synonym s-3-aminodihydrofuran-2 3h-one hydrobromide, s---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s-2-amino-4-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, s---alpha-amino-gamma-butyrolactonehydrobromide, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr
PubChem CID 16213138
SMILES Br.NC1CCOC1=O

Specifications

Melting Point 258°C to 260°C
Quantity 0.25 g
Beilstein 3912375
Sensitivity Hygroscopic
Optical Rotation −21° (c=1 in Water)
IUPAC Name 3-aminooxolan-2-one hydrobromide
Formula Weight 182.02
Percent Purity 98%
Chemical Name or Material L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.