missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-proline, TRC

Product Code. 30135677
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135677 1 g 1g
30132559 500 mg 500mg
17957744 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135677 Supplier Toronto Research Chemicals Supplier No. TRCA5113401G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13485-59-1
Synonym L-Proline, L-alanyl-∣L-Proline, 1-L-alanyl-∣Proline, 1-L-alanyl- (7CI)∣Proline, 1-L-alanyl-, L- (8CI)∣L-Alanyl-L-proline∣101: PN: US20070066537 PAGE: 17 claimed protein∣12: PN: WO2014170713 SEQID: 112 claimed protein∣150: PN: EP2161028 PAGE: 10 claimed protein∣443: PN: WO2005081628 SEQID: 445 claimed protein∣76: PN: WO2011146121 PAGE: 115 claimed sequence∣Alanylproline∣L-α-Alanyl-L-proline∣N-Alanylproline∣NSC 97932

Specifications

Format Neat
Molecular Weight (g/mol) 186.21
Product Type Impurity
Quantity 1 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.