missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-glutamine, TRC

Product Code. 30136968
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136968 5 g 5g
30133657 25 g 25g
17937654 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136968 Supplier Toronto Research Chemicals Supplier No. TRCA5113705G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39537-23-0
Molecular Formula C8 H15 N3 O4
Molecular Weight (g/mol) 217.22
Synonym L-Alanyl-L-Glutamine, L-Glutamine, L-alanyl-, Glutamine, N2-L-alanyl- (7CI), L-Glutamine, N2-L-alanyl-, L-Alanyl-L-glutamine, Alanyl-glutamine, Dipeptamin, Dipeptiven, GlutaMAX, Glutamax I, L-Ala-L-Gln, Sustamine
IUPAC Name (2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid
SMILES C[C@H](N)C(=O)N[C@@H](CCC(=O)N)C(=O)O

Specifications

Analyte or Component Names L-Alanyl-L-Glutamine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C8H15N3O4/c1-4(9)7(13)11-5(8(14)15)2-3-6(10)12/h4-5H,2-3,9H2,1H3,(H2,10,12)(H,11,13)(H,14,15)/t4-,5-/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 5 g
Formula Weight 217.1063
Chemical Name or Material L-Alanyl-L-Glutamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.