missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-glutamine, 99%, Thermo Scientific Chemicals

Catalog Number 15418589
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
39537-23-0
C8H15N3O4
217.23
MFCD00133046
HJCMDXDYPOUFDY-WHFBIAKZSA-N
l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
123935
CHEBI:73788
C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
This item is not returnable. View return policy
39537-23-0
C8H15N3O4
217.23
MFCD00133046
HJCMDXDYPOUFDY-WHFBIAKZSA-N
l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
123935
CHEBI:73788
C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O

L-Alanyl-L-glutamine, CAS # 39537-23-0, is a synthesized dipeptide, which can be digested to form stable L-glutamine in vitro.

A substitute for glutamine in mammalian cell culture media; stable to heat-sterilization.L-Alanyl-L-glutamine is used as a component in pharmaceuticals, cosmetics and food products. It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency. It provides nutritional requirements, improves intestinal health and enhances the immune system. It is also involved in various clinical studies such as cancer treatment, radiation and chemotherapy. It finds application in biotechnology as a component for mammalian cell culture medium. It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Alanyl-L-glutamine is a synthetic dipeptide that degrades to L-glutamine

Application

• L-Alanyl-L-glutamine can be used as a substitute for glutamine in mammalian cell culture media and is stable to heat-sterilization
• It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency
• It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release
• When this dipeptide is added to cultures of antibody-producing CHO cells in vitro, it reduces apoptosis and promotes antibody production

TRUSTED_SUSTAINABILITY
Melting Point 222°C
Boiling Point 615°C
Color White
Solubility Information Soluble in water.
IUPAC Name (2S)-2-[(2S)-2-aminopropanamido]-4-carbamoylbutanoic acid
Formula Weight 217.22
Percent Purity 99%
Quantity 5 g
Flash Point >110°C (230°F)
Physical Form Powder
Chemical Name or Material L-Alanyl-L-glutamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'