missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-glutamine, 99%

Product Code. p-7057579
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418589 5 g 5g
15428589 25 g 25g
15438589 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418589 Supplier Thermo Scientific Alfa Aesar Supplier No. J66996.06
Add to basket
This item is not returnable. View return policy

L-Alanyl-L-glutamine, CAS # 39537-23-0, is a synthesized dipeptide, which can be digested to form stable L-glutamine in vitro. | CAS: 39537-23-0 | C8H15N3O4 | 217.23 g/mol

A substitute for glutamine in mammalian cell culture media; stable to heat-sterilization.L-Alanyl-L-glutamine is used as a component in pharmaceuticals, cosmetics and food products. It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency. It provides nutritional requirements, improves intestinal health and enhances the immune system. It is also involved in various clinical studies such as cancer treatment, radiation and chemotherapy. It finds application in biotechnology as a component for mammalian cell culture medium. It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Alanyl-L-glutamine is a synthetic dipeptide that degrades to L-glutamine

Application

• L-Alanyl-L-glutamine can be used as a substitute for glutamine in mammalian cell culture media and is stable to heat-sterilization
• It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency
• It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release
• When this dipeptide is added to cultures of antibody-producing CHO cells in vitro, it reduces apoptosis and promotes antibody production

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39537-23-0
Molecular Formula C8H15N3O4
Molecular Weight (g/mol) 217.23
MDL Number MFCD00133046
InChI Key HJCMDXDYPOUFDY-WHFBIAKZSA-N
Synonym l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
PubChem CID 123935
ChEBI CHEBI:73788
SMILES C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 222°C
Boiling Point 615°C
Color White
Solubility Information Soluble in water.
IUPAC Name (2S)-2-[(2S)-2-aminopropanamido]-4-carbamoylbutanoic acid
Quantity 5 g
Flash Point >110°C (230°F)
Formula Weight 217.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Alanyl-L-glutamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.