missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine tert-butyl ester hydrochloride, 99%

Product Code. 11915401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11915401 1 g Glass Bottle 1g
11377651 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915401 Supplier Thermo Scientific Acros Supplier No. 442920010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13404-22-3 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13404-22-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00035524
InChI Key WIQIWPPQGWGVHD-JEDNCBNOSA-N
Synonym h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 11275390
SMILES Cl.C[C@H](N)C(=O)OC(C)(C)C

Specifications

Color White
Packaging Glass Bottle
IUPAC Name tert-butyl (2S)-2-aminopropanoate;hydrochloride
Quantity 1 g
Formula Weight 181.66
Physical Form Crystalline Powder or Solid
Assay 99%
Chemical Name or Material L-Alanine tert-butyl ester hydrochloride
Product Identifier
  • L-Alanine tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.