missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine methyl ester hydrochloride, 99%

Product Code. 11479472
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11479472

Brand: Thermo Scientific Alfa Aesar A10178.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2491-20-5 | C4H10ClNO2 | 139.58 g/mol

L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in Water (100 mg/ml).

Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-20-5
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00063663
InChI Key IYUKFAFDFHZKPI-DOIAFFBONA-N
Synonym l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
PubChem CID 2733257
IUPAC Name methyl (2S)-2-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](C)N

Specifications

Melting Point 106°C to 110°C
Beilstein 3594033
Solubility Information Soluble in Water (100mg/ml).
Quantity 5 g
Formula Weight 139.58
Optical Rotation +6.5° (c=10 in methanol)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Alanine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.