missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine methyl ester hydrochloride, 99%

Product Code. 11489472
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489472 25 g 25g
11479472 5 g 5g
11499472 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489472 Supplier Thermo Scientific Alfa Aesar Supplier No. A10178.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2491-20-5 | C4H10ClNO2 | 139.58 g/mol

L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in Water (100 mg/ml).

Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-20-5
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00063663
InChI Key IYUKFAFDFHZKPI-DOIAFFBONA-N
Synonym l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
PubChem CID 2733257
IUPAC Name methyl (2S)-2-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](C)N

Specifications

Melting Point 106°C to 110°C
Beilstein 3594033
Solubility Information Soluble in Water (100mg/ml).
Quantity 25 g
Formula Weight 139.58
Optical Rotation +6.5° (c=10 in methanol)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Alanine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.