missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine isopropyl ester hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 167.63 |
| Percent Purity | 95% |
| Chemical Name or Material | L-Alanine isopropyl ester hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction