missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine ethyl ester hydrochloride, 98+%

Product Code. 11493624
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493624 25 g 25g
11483624 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493624 Supplier Thermo Scientific Alfa Aesar Supplier No. A11271.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1115-59-9 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-59-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00063662
InChI Key JCXLZWMDXJFOOI-UHFFFAOYNA-N
Synonym l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride
PubChem CID 2724356
IUPAC Name ethyl (2S)-2-aminopropanoate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)C(C)N

Specifications

Melting Point 74°C to 78°C
Beilstein 3594395
Quantity 25 g
Formula Weight 153.61
Optical Rotation +2.9° (c=2.5 in water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Alanine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.