missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine-d3, TRC

Product Code. 30179498
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179498 50 mg 50mg
30179129 100 mg 100mg
30179107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179498 Supplier Toronto Research Chemicals Supplier No. TRCA48150350MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63546-27-0
Molecular Formula C3 D3 H4 N O2
Molecular Weight (g/mol) 92.112
Synonym L-Alanine-3,3,3-d3 (9CI, ACI), L-[3,3,3-d3]Alanine, β,β,β-Trideutero-L-alanine, (2S)-2-Aminopropanoic-3,3,3-d3 acid, L-Alanine-3,3,3-D3, L-Alanine-d3
IUPAC Name (2S)-2-amino-3,3,3-trideuteriopropanoic acid
SMILES [2H]C([2H])([2H])[C@H](N)C(=O)O

Spezifikation

Quantity 50 mg
InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i1D3
Formula Weight 92.067
Chemical Name or Material L-Alanine-d3
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.