missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine benzyl ester hydrochloride, 98%

Product Code. 15442609
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442609 25 g 25g
15432609 5 g 5g
15452609 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442609 Supplier Thermo Scientific Alfa Aesar Supplier No. H65699.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5557-83-5 | C10H14ClNO2 | 215.677 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5557-83-5
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.677
MDL Number MFCD00054340
InChI Key RLMHWGDKMJIEHH-QRPNPIFTSA-N
Synonym h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
PubChem CID 12210893
IUPAC Name benzyl (2S)-2-aminopropanoate;hydrochloride
SMILES CC(C(=O)OCC1=CC=CC=C1)N.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 215.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Alanine benzyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.