missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, 99%, MP Biomedicals™

Product Code. 11452970 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Unit Size:
1kg
25g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452970 5 kg 5kg
11412970 25 g 25g
11442970 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452970 Supplier MP Biomedicals Supplier No. 0210028705
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 0.99
Density 1.40g/mL
Quantity 5 kg
Formula Weight 89.1
Percent Purity ≥99%
Grade Research
Physical Form Powder
Chemical Name or Material L-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.