missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, 99%, MP Biomedicals™

Product Code. 11412970 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Unit Size:
1kg
25g
5kg
Product Code. Quantity unitSize
11412970 25 g 25g
11442970 1 kg 1kg
11452970 5 kg 5kg
3 options
This item is not returnable. View return policy

Product Code. 11412970

Brand: MP Biomedicals 02100287.3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 0.99
Density 1.40g/mL
Quantity 25 g
Formula Weight 89.1
Physical Form Powder
Chemical Name or Material L-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.