missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanine, 99%

Product Code. p-7022853
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11442468

Brand: Thermo Scientific Chemicals A15804.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol

L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Melting Point ∽295°C (decomposition)
Assay Percent Range 99%
Beilstein 1720248
Merck Index 14,204
Solubility Information Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone.
Density 1.432
Quantity 25 g
Formula Weight 89.09
Percent Purity 99%
Chemical Name or Material L-Alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.