missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine 4-nitroanilide hydrochloride, 98%

Product Code. 15490059
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490059 1 g 1g
15410069 250 mg 250mg
15400069 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490059 Supplier Thermo Scientific Alfa Aesar Supplier No. H63393.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31796-55-1 | C9H12ClN3O3 | 245.663 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31796-55-1
Molecular Formula C9H12ClN3O3
Molecular Weight (g/mol) 245.663
MDL Number MFCD00039088
InChI Key YEXRLSXNWLNHQR-RGMNGODLSA-N
Synonym h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864
PubChem CID 2802426
IUPAC Name (2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride
SMILES CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl

Specifications

Color Yellow
Beilstein 6303731
Quantity 1 g
Formula Weight 245.66
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Alanine 4-nitroanilide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.