missing translation for 'onlineSavingsMsg'
Learn More

L-Adrenaline, Mikromol™

Product Code. 18012070
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18012070 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18012070 Supplier Mikromol Supplier No. MM0614.02
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Adrenaline
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Adrenaline
CAS 51-43-4
Analyte or Component Names L-Adrenaline
Shipping Condition Room Temperature
Molecular Formula C9 H13 N O3
Certifications/Compliance ISO 17034
Synonym (-)-Epinephrine, Noradrenaline Tartrate Imp. A (EP), Noradrenaline Imp. A (EP), 1,2-Benzenediol, 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]-, 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, (R)-, Benzyl alcohol, 3,4-dihydroxy-alpha-[(methylamino)methyl]-, (-)- (8CI), 4-[(1R)-1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol, (-)-(R)-Epinephrine, (-)-3,4-Dihydroxy-alpha-[2-(methylamino)ethyl]benzyl alcohol, (-)-Adrenaline, (R)-Adrenaline, (R)-Epinephrine, Adnephrine, Adrenal, Adrenalin, Adrenaline, Adrenine, Adrin, Ana-Kit, Bosmin, Bronkaid Mist, Chelafrin, Epifrin, Epiglaufrin, Epinefrina, Epinephran, Epinephrine, Epipen, Epirenan, Eppy, Exadrin, Glauposine, Hemisine, Hemostasin, Hemostatin, Hypernephrin, Isoptoepinal, L-Adrenaline, L-Methylaminoethanolcatechol, Levoepinephrine, Levorenen, Levorenin, Levorenine, Levoreninum, Lyodrin, Methylarterenol, Mucidrina, N-[(2R)-2-(3,4-Dihydroxyphenyl)-2-hydroxyethyl]-N-methylamine, NSC 62786, Nephridine, Nieraline, Paranephrin, Primatene Mist, R-(-)-Epinephrine, Renaglandin, Renaleptine, Renalina, Renoform, Renostypticin, Renostyptin, Scurenaline, Simplene, Styptirenal, Supracapsulin, Supranephrane, Suprarenaline, Suprarenin, Surrenine, Sus-phrine, Takamina, Vasoconstrictine, Vasotonin, l-1-(3,4-Dihydroxyphenyl)-2-methylaminoethanol, l-Adrenaline, l-Epinephrine, l-Epirenamine, Noradrenaline Tartrate Impurity A, Noradrenaline Impurity A
InChI Formula InChI=1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/t9-/m0/s1
SMILES CNC[C@H](O)c1ccc(O)c(O)c1
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Molecular Weight (g/mol) 183.2
Quantity 500 mg
Formula Weight 183.0895 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.