missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Abrine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | L-Abrine |
| CAS | 526-31-8 |
| Molecular Formula | C12 H14 N2 O2 |
| Quantity | 250 mg |
| Synonym | L-Tryptophan, N-methyl-, Tryptophan, N-methyl-, L- (6CI,7CI,8CI), N-Methyl-L-tryptophan, (2S)-3-(1H-Indol-3-yl)-2-(methylazaniumyl)propanoate, (S)-3-(1H-Indol-3-yl)-2-methylaminopropionic acid, (2S)-3-(1H-Indol-3-yl)-2-methylaminopropanoic acid, Abrine, Abrine (amino acid), L-(+)-Abrine, L-Abrine, L-N-Methyltryptophan, N-Methyltryptophan, N-α-Methyl-L-tryptophan, NPLC 0233 |
| InChI Formula | InChI=1S/C12H14N2O2/c1-13-11(12(15)16)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,11,13-14H,6H2,1H3,(H,15,16)/t11-/m0/s1 |
| SMILES | CN[C@@H](Cc1c[nH]c2ccccc12)C(=O)O |
| IUPAC Name | (2S)-3-(1H-indol-3-yl)-2-(methylamino)propanoic acid |
| Molecular Weight (g/mol) | 218.2518 |
| Formula Weight | 218.1055 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction