missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-838,417, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Subtype-selective GABAA receptor partial agonist. Selectively binds to α1, α2, α3 and α5 subunits (Ki values are 0.79, 0.67, 0.67 and 2.25 nM respectively) but displays no efficacy at α1 (α1-sparing). Exhibits non-sedative anxiolytic, antinociceptive and anti-inflammatory activity in vivo.
Specifications
Quantity | 10 mg |
Synonym | unii-8czo0970g3, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy-1,2,4-triazolo 4,3-b pyridazine, 7-tert-butyl-3-2,5-difluorophenyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy-1,2,4 triazolo 4,3-b pyridazine, 5-7-tert-butyl-3-2,5-difluorophenyl-1,2,4 triazolo 4,3-b pyridazin-6-yl oxy methyl-1-methyl-1,2,4-triazole, 1,2,4-triazolo 4,3-b pyridazine, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy, l hplc, 1,2,4-triazolo 4,3-b pyridazine,3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4-triazol-5-yl methoxy, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-2-methyl-2h-1,2,4-triazol-3-ylmethoxy-1,2,4-triazolo 4,3-b pyridazine, 3-2,5-difluorophenyl-7-1,1-dimethylethyl-6-1-methyl-1h-1,2,4,-triazol-5-yl methoxy-1,2,4-triazolo 4,3-b pyridazine |
Formula Weight | 399.4 |
Percent Purity | >98% |
Chemical Name or Material | L-838, 417 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'