missing translation for 'onlineSavingsMsg'
Learn More

L-778123 hydrochloride, MedChemExpress

Product Code. 16477076
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477076 100 mg 100mg
16473067 50 mg 50mg
16467018 10 mg 10mg
16409567 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477076 Supplier Medchem Express Supplier No. HY16273A
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-778123 hydrochloride is an inhibitor of FPTase and GGPTase-I with IC50 of 2 nM and 98 nM in enzyme inhibition determination.

L-778123 hydrochloride is an inhibitor of FPTase and GGPTase-I with IC50 of 2 nM and 98 nM in enzyme inhibition determination.

Specifications

Chemical Name or Material L-778123 hydrochloride
CAS 253863-00-2
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl2N5O
Quantity 100 mg
Synonym L-778,123 hydrochloride
Solubility Information DMSO : ≥ 25 mg/mL (56.52 mM) ∣H2O : 25 mg/mL (56.52 mM; Need ultrasonic)
SMILES N#CC1=CC=C(CN2C(CN3CC(N(C4=CC=CC(Cl)=C4)CC3)=O)=CN=C2)C=C1.[H]Cl
Molecular Weight (g/mol) 442.34
Formula Weight 442.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.