Learn More
L-745,870 trihydrochloride, Tocris Bioscience™
Catalog Number 13448263
Encompass
Click to view available options
10mg
Highly selective D4 antagonist
A highly potent and selective D4 dopamine receptor antagonist; L-745,870 has Ki values of 0.51, 2300 and 960 nM for D4, D3 and D2 subtypes respectively and > 1000-fold selectivity over 5-HT2, D1 and D5 receptors.