missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-741,742 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A potent and highly selective D4 dopamine receptor antagonist, with Ki values of > 1700, 770 and 3.5 nM at cloned human D2, D3 and D4 receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 417.38 |
| Purity | >98% |
| Molecular Weight (g/mol) | 417.374 |
| Chemical Name or Material | L-741, 742 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction