missing translation for 'onlineSavingsMsg'
Learn More

L-741,626, Tocris Bioscience™

Catalog Number p-4783575
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
81226-60-0
C20H21ClN2O
340.851
LLBLNMUONVVVPG-UHFFFAOYSA-N
3-4-4-chlorophenyl-4-hydroxypiperidino methyl indole, 4-4-chlorophenyl-1-1h-indol-3-ylmethyl piperidin-4-ol, 4-4-chlorophenyl-1-1h-indol-3-ylmethyl-4-piperidinol, 4-piperidinol, 4-4-chlorophenyl-1-1h-indol-3-ylmethyl, tocris-1003, lopac-l-135, biomol-nt_000036, d0m3pv, gtpl177, l hplc
133633
4-(4-chlorophenyl)-1-(1H-indol-3-ylmethyl)piperidin-4-ol
C1CN(CCC1(C2=CC=C(C=C2)Cl)O)CC3=CNC4=CC=CC=C43
This item is not returnable. View return policy
81226-60-0
C20H21ClN2O
340.851
LLBLNMUONVVVPG-UHFFFAOYSA-N
3-4-4-chlorophenyl-4-hydroxypiperidino methyl indole, 4-4-chlorophenyl-1-1h-indol-3-ylmethyl piperidin-4-ol, 4-4-chlorophenyl-1-1h-indol-3-ylmethyl-4-piperidinol, 4-piperidinol, 4-4-chlorophenyl-1-1h-indol-3-ylmethyl, tocris-1003, lopac-l-135, biomol-nt_000036, d0m3pv, gtpl177, l hplc
133633
4-(4-chlorophenyl)-1-(1H-indol-3-ylmethyl)piperidin-4-ol
C1CN(CCC1(C2=CC=C(C=C2)Cl)O)CC3=CNC4=CC=CC=C43

High affinity D2 antagonist

A potent D2 dopamine receptor selective antagonist, with affinities of 2.4, 100 and 220 nM for D2, D3 and D4 receptors respectively. Centrally active following systemic administration in vivo.
Quantity 50 mg
Formula Weight 340.85
Percent Purity >98%
Chemical Name or Material L-741, 626
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'