missing translation for 'onlineSavingsMsg'
Learn More

L-701,252, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001414
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001414

Brand: Tocris Bioscience 0705/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

NMDA antagonist, acts glycine site

An antagonist at the glycine-NMDA site (IC50 = 420 nM). Also a potent systemic anticonvulsant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151057-13-5
Molecular Formula C13H10ClNO3
Molecular Weight (g/mol) 263.677
InChI Key MXEFWCFPCLDOOG-UHFFFAOYSA-N
Synonym 7-chloro-3-cyclopropanecarbonyl-4-hydroxyquinolin-2 1h-one, 7-chloro-3-cyclopropyl-carbonyl-4-hydroxy-2 1h-quinolinone, 7-chloro-3-cyclopropylcarbonyl-4-hydroxy-2 1h-quinolinone, tocris-0705, biomol-nt_000184, 2 1h-quinolinone,7-chloro-3-cyclopropylcarbonyl-4-hydroxy, 7-chloranyl-3-cyclopropylcarbonyl-2-oxidanyl-1h-quinolin-4-one, 7-chloro-3-cyclopropanecarbonyl-2-hydroxy-1h-quinolin-4-one, 7-chloro-3-cyclopropyl-carbonyl-4-hydroxy-2 1h quinolinone, 7-chloro-3-cyclopropylcarbonyl-4-hydroxy-2 1h-quinoline
PubChem CID 54687453
IUPAC Name 7-chloro-3-(cyclopropanecarbonyl)-4-hydroxy-1H-quinolin-2-one
SMILES C1CC1C(=O)C2=C(C3=C(C=C(C=C3)Cl)NC2=O)O

Specifications

Quantity 10 mg
Formula Weight 263.68
Percent Purity >99%
Chemical Name or Material L-701, 252
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.