missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-689,560

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
L-689,560 is a very potent antagonist at the glycine-NMDA site.
Specifications
Specifications
| Chemical Name or Material | trans-2-Carboxy-5,7-dichloro-4-phenylaminocarbonylamino-1,2,3,4-tetrahydroquinoline |
| CAS | 139051-78-8 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Solubility | Soluble to 25mM in DMSO |
| Target | NMDA Receptor Antagonists |
| Molecular Formula | C17H15Cl2N3O3 |
| Formula Weight | Observed MW: 380.23 |
| Purity | 99% |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction