missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-689,560
GREENER_CHOICE

Product Code. 18055392
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055392 50 mg 50mg
18707636 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18055392 Supplier Tocris Bioscience™ Supplier No. 0742/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent NMDA antagonist

L-689,560 is a very potent antagonist at the glycine-NMDA site.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-2-Carboxy-5,7-dichloro-4-phenylaminocarbonylamino-1,2,3,4-tetrahydroquinoline
CAS 139051-78-8
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 25mM in DMSO
Target NMDA Receptor Antagonists
Molecular Formula C17H15Cl2N3O3
Formula Weight Observed MW: 380.23
Purity 99%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.