missing translation for 'onlineSavingsMsg'
Learn More

L-685458, MedChemExpress

Product Code. 16450218
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16450218

Brand: Medchem Express HY19369

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-685458 is a potent transition state analog (TSA) γ-secretase inhibitor (GSI).

L-685458 inhibits amyloid β-protein precursor γ-secretase activity with IC50 of 17 nM, shows greater than 50-100-fold selectivity over other aspartyl proteases tested. L685458 inhibits γ-secretase-mediated cleavage of APP-C99 and Notch-100 with IC50s of 301.3 nM and 351.3 nM, respectively. L-685458 can be used for the research of alzheimer’s disease (AD) and cancers.

Specifications

Chemical Name or Material L-685458
Physical Form Solid
CAS 292632-98-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C39H52N4O6
For Use With (Application) Cancer-programmed cell death
Synonym L-685,458
Solubility Information DMSO : 83.3 mg/mL (123.80 mM; Need ultrasonic and warming)
SMILES O=C(N)[C@H](CC1=CC=CC=C1)NC([C@H](CC(C)C)NC([C@H](CC2=CC=CC=C2)C[C@@H](O)[C@@H](NC(OC(C)(C)C)=O)CC3=CC=CC=C3)=O)=O
Molecular Weight (g/mol) 672.85
Formula Weight 672.85
Percent Purity 99.09%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.