missing translation for 'onlineSavingsMsg'
Learn More

L-5-Hydroxytryptophan, MedChemExpress

Product Code. 16418806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418806 Supplier Medchem Express Supplier No. HYB1716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-5-Hydroxytryptophan (L-5-HTP), a naturally occurring amino acid and a dietary supplement for use as an antidepressant, appetite suppressant, and sleep aid, is the immediate precursor of the neurotransmitter serotonin and a reserpine antagonist. L-5-Hydroxytryptophan (L-5-HTP) is used to treat fibromyalgia, myoclonus, migraine, and cerebellar ataxia.

L-5-Hydroxytryptophan (L-5-HTP), a naturally occurring amino acid and a dietary supplement for use as an antidepressant, appetite suppressant, and sleep aid, is the immediate precursor of the neurotransmitter serotonin and a reserpine antagonist. L-5-Hydroxytryptophan (L-5-HTP) is used to treat fibromyalgia, myoclonus, migraine, and cerebellar ataxia.

Specifications

Chemical Name or Material L-5-Hydroxytryptophan
CAS 9-8-4350
Color Tan
Purity Grade Notes Research
Molecular Formula C11H12N2O3
Synonym L-5-HTP Oxitriptan
Solubility Information DMSO : 12.5 mg/mL (56.76 mM; ultrasonic and warming and heat to 60°C)
SMILES N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O
Molecular Weight (g/mol) 220.22
Quantity 100 mg
Formula Weight 220.22
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.