missing translation for 'onlineSavingsMsg'
Learn More

L-5-Hydroxytryptophan hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11451698
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451698 1 g 1g
11461698 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451698 Supplier Thermo Scientific Alfa Aesar Supplier No. A13954.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-8-4350
MDL Number MFCD00064341
ChEBI CHEBI:17780

Specifications

Melting Point 258°C (decomposition)
Molecular Formula C11H12N2O3
UN Number UN2811
Beilstein 88200
Merck Index 14,4847
Synonym 5-hydroxy-l-tryptophan, l-5-hydroxytryptophan, oxitriptan, 5-hydroxytryptophan, levothym, tript-oh, cincofarm, l-5-htp, pretonine, 2s-2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid
InChI Key LDCYZAJDBXYCGN-IMWMWJONNA-N
SMILES N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O
IUPAC Name (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 220.23
Quantity 1 g
PubChem CID 439280
Formula Weight 220.23 (Anhydrous)
Optical Rotation −33° (c=1 in Water)
Percent Purity 98%
Chemical Name or Material L-5-Hydroxytryptophan hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.