missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-5-Hydroxytryptophan, 99%

Catalog Number 10143480
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
C11H12N2O3
MFCD00064341
This item is not returnable. View return policy
C11H12N2O3
MFCD00064341

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Arsenic (As) 1ppm max.
Color Beige to White or Gray
Heavy Metals 10ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Merck Index 15, 4886
Packaging Glass Bottle
Specific Rotation -30.9° to -34.9° (20°C, 589nm) (c=1, H2O) on dry substance
Formula Weight 220.23
Percent Purity 98.5 to 101.5% (on dry substance)
Sulfated Ash 0.3% max.
Water 1% max. (K.F.)
Specific Rotation Condition - 32.50 (20.00°C c=1,H2O on dry basis)
Quantity 5 g
Physical Form Powder
Chemical Name or Material L-5-Hydroxytryptophan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'